Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nemy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169657.2 | NP_001163128.1 | 4 - 5 | 793 | 61 | |
NM_001299429.1 | NP_001286358.1 | 3 - 4 | 1260 | 61 | ||
NM_001299430.1 | NP_001286359.1 | 3 - 4 | 935 | 61 | ||
NM_136964.4 | NP_610808.1 | 3 - 4 | 1260 | 61 | ||
NM_165934.3 | NP_725208.1 | 4 - 5 | 938 | 61 | ||
NM_165935.3 | NP_725209.1 | 4 - 5 | 748 | 61 | ||
NM_165936.3 | NP_725210.1 | 3 - 4 | 545 | 61 | ||
NM_165937.3 | NP_725211.3 | 3 - 4 | 743 | 61 | ||
GenBank mRNA | AY060294.1 | - | 3 - 4 | 1246 | 61 | |
BT003222.1 | - | 4 - 5 | 936 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |