Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Indy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169994.2 | NP_001163465.1 | 8 - 9 | 1700 | 92 | |
NM_079426.4 | NP_524150.1 | 8 - 9 | 1603 | 92 | ||
NM_168778.2 | NP_730363.2 | 8 - 9 | 1691 | 92 | ||
NM_168779.2 | NP_730364.1 | 8 - 9 | 1719 | 92 | ||
GenBank mRNA | AF509505.1 | - | 8 - 9 | 1686 | 92 | |
AY102686.1 | - | 8 - 9 | 1567 | 92 | ||
BT100230.1 | - | 8 - 9 | 1685 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |