Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275175.1 | NP_001262104.1 | 2 - 3 | 292 | 110 | |
NM_001275176.1 | NP_001262105.1 | 2 - 3 | 292 | 110 | ||
NM_140953.3 | NP_649210.1 | 1 - 2 | 848 | 110 | ||
NM_168845.2 | NP_730517.1 | 2 - 3 | 328 | 110 | ||
NM_168846.2 | NP_730518.1 | 2 - 3 | 292 | 110 | ||
NM_168847.2 | NP_730519.1 | 2 - 3 | 557 | 110 | ||
NM_168848.2 | NP_730520.1 | 2 - 3 | 315 | 110 | ||
NM_176369.3 | NP_788547.1 | 1 - 2 | 266 | 110 | ||
GenBank mRNA | AY069511.1 | - | 1 - 2 | 293 | 110 | |
BT120009.1 | - | 2 - 3 | 299 | 110 | ||
BT120010.1 | - | 2 - 3 | 302 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Drosophila melanogaster |