Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coro | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273813.1 | NP_001260742.1 | 5 - 6 | 1143 | 110 | |
NM_001273814.1 | NP_001260743.1 | 4 - 5 | 1169 | 110 | ||
NM_001273815.1 | NP_001260744.1 | 4 - 5 | 1008 | 110 | ||
NM_001273816.1 | NP_001260745.1 | 4 - 5 | 983 | 110 | ||
NM_136398.4 | NP_610242.1 | 4 - 5 | 1125 | 110 | ||
NM_165494.2 | NP_724509.1 | 4 - 5 | 1025 | 110 | ||
NM_206031.2 | NP_995753.1 | 4 - 5 | 1104 | 110 | ||
GenBank mRNA | AF467271.1 | - | 4 - 5 | 1086 | 110 | |
AY118560.1 | - | 4 - 5 | 1176 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |