Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014503.2 | NP_001014503.1 | 10 - 11 | 3563 | 78 | |
NM_001169610.2 | NP_001163081.1 | 11 - 12 | 3556 | 78 | ||
NM_001169611.1 | NP_001163082.1 | 11 - 12 | 3580 | 78 | ||
NM_001169612.2 | NP_001163083.1 | 12 - 13 | 3622 | 78 | ||
NM_001259270.2 | NP_001246199.1 | 11 - 12 | 3556 | 78 | ||
NM_001259271.2 | NP_001246200.1 | 11 - 12 | 3460 | 78 | ||
NM_001273865.1 | NP_001260794.1 | 10 - 11 | 3624 | 78 | ||
NM_001273866.2 | NP_001260795.1 | 9 - 10 | 3343 | 78 | ||
NM_001316409.1 | NP_001303338.1 | 9 - 10 | 3343 | 78 | ||
NM_136504.5 | NP_610348.3 | 11 - 12 | 3556 | 78 | ||
NM_165586.5 | NP_724643.2 | 11 - 12 | 3625 | 78 | ||
NM_206056.5 | NP_995778.2 | 11 - 12 | 3911 | 78 | ||
GenBank mRNA | AF276964.1 | - | 10 - 11 | 3563 | 78 | |
AY069682.1 | - | 11 - 12 | 3580 | 78 | ||
BT100315.1 | - | 10 - 11 | 3590 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |