Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165735.4 | NP_724878.2 | 6 - 7 | 1527 | 94 | |
NM_206069.3 | NP_995791.1 | 6 - 7 | 1509 | 94 | ||
GenBank mRNA | AB073865.1 | - | 6 - 7 | 1289 | 94 | |
AF149799.1 | - | 6 - 7 | 1487 | 94 | ||
AF521176.1 | - | 6 - 7 | 1521 | 94 | ||
AY115551.1 | - | 6 - 7 | 881 | 94 | ||
BT021371.1 | - | 6 - 7 | 1487 | 94 | ||
BT072996.1 | - | 6 - 7 | 1510 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |