Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136731.2 | NP_610575.1 | 3 - 4 | 1853 | 58 | |
NM_206074.2 | NP_995796.2 | 3 - 4 | 1853 | 58 | ||
GenBank mRNA | BT031149.1 | - | 3 - 4 | 1826 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |