Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136731.2 | NP_610575.1 | 8 - 9 | 3468 | 73 | |
NM_206074.2 | NP_995796.2 | 8 - 9 | 3468 | 73 | ||
GenBank mRNA | BT031149.1 | - | 8 - 9 | 3441 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |