Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014520.3 | NP_001014520.2 | 4 - 5 | 2050 | 73 | |
NM_001316414.1 | NP_001303343.1 | 4 - 5 | 2050 | 73 | ||
NM_001382049.1 | NP_001369078.1 | 4 - 5 | 2050 | 73 | ||
NM_001382050.1 | NP_001369079.1 | 2 - 3 | 1408 | 73 | ||
NM_001382051.1 | NP_001369080.1 | 2 - 3 | 1408 | 73 | ||
NM_078962.4 | NP_523686.2 | 4 - 5 | 2050 | 73 | ||
NM_165790.3 | NP_724955.1 | 4 - 5 | 2050 | 73 | ||
NM_165791.3 | NP_724958.1 | 2 - 3 | 1408 | 73 | ||
NM_165792.3 | NP_724959.1 | 3 - 4 | 1147 | 73 | ||
NM_176141.3 | NP_788321.1 | 3 - 4 | 1463 | 73 | ||
NM_176142.3 | NP_788322.1 | 3 - 4 | 1196 | 73 | ||
NM_176143.3 | NP_788323.1 | 2 - 3 | 1233 | 73 | ||
NM_206086.3 | NP_995808.1 | 4 - 5 | 1893 | 73 | ||
GenBank mRNA | AY069588.1 | - | 4 - 5 | 1971 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |