Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169707.2 | NP_001163178.1 | 4 - 5 | 1481 | 59 | |
NM_137400.4 | NP_611244.1 | 4 - 5 | 1392 | 59 | ||
NM_206157.2 | NP_995879.1 | 4 - 5 | 1385 | 59 | ||
GenBank mRNA | AF181625.1 | - | 4 - 5 | 1375 | 59 | |
BT083389.1 | - | 4 - 5 | 1468 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |