Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299840.1 | NP_001286769.1 | 8 - 9 | 2105 | 69 | |
NM_079100.3 | NP_523824.1 | 8 - 9 | 2105 | 69 | ||
GenBank mRNA | M20630.1 | - | 8 - 9 | 2111 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |