Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gug | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259741.1 | NP_001246670.1 | 11 - 12 | 6399 | 56 | |
NM_001259742.1 | NP_001246671.1 | 11 - 12 | 6762 | 56 | ||
NM_001259743.1 | NP_001246672.1 | 11 - 12 | 6471 | 56 | ||
NM_001274651.1 | NP_001261580.1 | 11 - 12 | 6747 | 56 | ||
NM_001274652.1 | NP_001261581.1 | 12 - 13 | 6819 | 56 | ||
NM_079249.3 | NP_523973.2 | 12 - 13 | 6762 | 56 | ||
NM_145106.1 | NP_659574.1 | 11 - 12 | 6209 | 56 | ||
NM_176300.1 | NP_788478.1 | 12 - 13 | 6266 | 56 | ||
NM_206303.2 | NP_996025.1 | 12 - 13 | 6753 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |