Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sytalpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201903.1 | NP_001188832.1 | 2 - 3 | 279 | 56 | |
NM_001201904.1 | NP_001188833.1 | 2 - 3 | 537 | 56 | ||
NM_001299090.1 | NP_001286019.1 | 2 - 3 | 537 | 56 | ||
NM_136009.2 | NP_609853.1 | 2 - 3 | 385 | 56 | ||
GenBank mRNA | BT060450.1 | - | 2 - 3 | 279 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |