Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299141.1 | NP_001286070.1 | 2 - 3 | 269 | 59 | |
NM_136075.3 | NP_609919.1 | 2 - 3 | 269 | 59 | ||
GenBank mRNA | BT024445.1 | - | 2 - 3 | 210 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |