Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACXA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259082.1 | NP_001246011.1 | 7 - 8 | 2107 | 127 | |
NM_001259083.1 | NP_001246012.1 | 7 - 8 | 2102 | 127 | ||
NM_001273494.1 | NP_001260423.1 | 7 - 8 | 2107 | 127 | ||
NM_001298954.1 | NP_001285883.1 | 7 - 8 | 2040 | 127 | ||
NM_138990.2 | NP_620475.2 | 7 - 8 | 2107 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |