Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103834.4 | NP_001097304.2 | 10 - 11 | 2442 | 83 | |
NM_001103835.2 | NP_001097305.1 | 10 - 11 | 2418 | 83 | ||
NM_001103836.4 | NP_001097306.2 | 10 - 11 | 2478 | 83 | ||
NM_001144194.2 | NP_001137666.1 | 10 - 11 | 2445 | 83 | ||
NM_001299479.1 | NP_001286408.1 | 9 - 10 | 2121 | 83 | ||
NM_001299480.1 | NP_001286409.1 | 10 - 11 | 2466 | 83 | ||
NM_001316419.1 | NP_001303348.1 | 9 - 10 | 2166 | 83 | ||
NM_079014.3 | NP_523738.2 | 9 - 10 | 2139 | 83 | ||
NM_166034.2 | NP_725362.1 | 10 - 11 | 2442 | 83 | ||
NM_166035.2 | NP_725363.1 | 10 - 11 | 1962 | 83 | ||
NM_166036.2 | NP_725364.1 | 10 - 11 | 1947 | 83 | ||
GenBank mRNA | AF276664.1 | - | 9 - 10 | 2158 | 83 | |
BT023511.1 | - | 9 - 10 | 2154 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |