Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259396.2 | NP_001246325.1 | 2 - 3 | 388 | 77 | |
NM_058126.5 | NP_477474.1 | 1 - 2 | 553 | 77 | ||
NM_166065.3 | NP_725412.2 | 2 - 3 | 440 | 77 | ||
GenBank mRNA | AF145309.1 | - | 1 - 2 | 315 | 77 | |
AF160939.1 | - | 1 - 2 | 523 | 77 | ||
BT089015.1 | - | 2 - 3 | 420 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |