Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144457.3 | NP_001137929.1 | 2 - 3 | 797 | 100 | |
NM_140096.5 | NP_648353.3 | 2 - 3 | 797 | 100 | ||
NM_206310.3 | NP_996032.1 | 2 - 3 | 797 | 100 | ||
GenBank mRNA | AY060437.1 | - | 2 - 3 | 762 | 100 | |
BT001491.1 | - | 2 - 3 | 714 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |