Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078613.4 | NP_511168.3 | 6 - 7 | 1049 | 63 | |
NM_206724.2 | NP_996447.1 | 6 - 7 | 1204 | 63 | ||
GenBank mRNA | AF132025.1 | - | 6 - 7 | 1053 | 63 | |
BT021401.1 | - | 6 - 7 | 1053 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |