Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144728.3 | NP_001138200.1 | 16 - 17 | 2904 | 66 | |
NM_001258775.3 | NP_001245704.1 | 16 - 17 | 2904 | 66 | ||
NM_001258776.2 | NP_001245705.1 | 17 - 18 | 2895 | 66 | ||
NM_001272661.2 | NP_001259590.1 | 17 - 18 | 3103 | 66 | ||
NM_001272662.2 | NP_001259591.1 | 17 - 18 | 3103 | 66 | ||
NM_001298382.1 | NP_001285311.1 | 16 - 17 | 3586 | 66 | ||
NM_078634.7 | NP_523358.5 | 16 - 17 | 3586 | 66 | ||
NM_206752.3 | NP_996475.2 | 16 - 17 | 2904 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |