Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134691.2 | NP_608535.1 | 1 | 1073 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |