Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eys | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032399.3 | NP_001027571.3 | 12 - 13 | 6724 | 86 | |
NM_001272995.1 | NP_001259924.2 | 12 - 13 | 6724 | 86 | ||
GenBank mRNA | DQ991915.1 | - | 12 - 13 | 6711 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |