Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Art2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298671.1 | NP_001285600.1 | 2 - 3 | 387 | 59 | |
NM_134977.2 | NP_608821.1 | 1 - 2 | 228 | 59 | ||
GenBank mRNA | BT024987.1 | - | 1 - 2 | 228 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |