Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: teq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169922.3 | NP_001163393.2 | 17 - 18 | 7059 | 86 | |
NM_140031.3 | NP_648288.2 | 16 - 17 | 8298 | 86 | ||
NM_168314.3 | NP_729451.1 | 7 - 8 | 4272 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |