Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: teq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169922.3 | NP_001163393.2 | 18 - 19 | 7138 | 61 | |
NM_140031.3 | NP_648288.2 | 17 - 18 | 8377 | 61 | ||
NM_168314.3 | NP_729451.1 | 8 - 9 | 4351 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |