Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: teq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169922.3 | NP_001163393.2 | 7 - 8 | 2221 | 81 | |
NM_140031.3 | NP_648288.2 | 7 - 8 | 2221 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |