Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169928.2 | NP_001163399.1 | 4 - 5 | 943 | 86 | |
NM_001169929.2 | NP_001163400.1 | 4 - 5 | 923 | 86 | ||
NM_140043.5 | NP_648300.3 | 5 - 6 | 929 | 86 | ||
NM_168325.4 | NP_729469.2 | 5 - 6 | 949 | 86 | ||
GenBank mRNA | AY069422.1 | - | 5 - 6 | 805 | 86 | |
BT003777.1 | - | 5 - 6 | 864 | 86 | ||
BT125863.1 | - | 4 - 5 | 835 | 86 | ||
BT133238.1 | - | 4 - 5 | 821 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |