Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nudE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140116.3 | NP_648373.1 | 5 - 6 | 1164 | 92 | |
GenBank mRNA | AY061280.1 | - | 5 - 6 | 1147 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |