Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140298.3 | NP_648555.1 | 1 - 2 | 282 | 77 | |
GenBank mRNA | AY095036.1 | - | 1 - 2 | 248 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |