Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sti | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274839.1 | NP_001261768.1 | 1 - 2 | 434 | 58 | |
NM_140327.2 | NP_648584.1 | 1 - 2 | 434 | 58 | ||
GenBank mRNA | BT081968.1 | - | 1 - 2 | 388 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |