Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140368.2 | NP_648625.1 | 1 - 2 | 747 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |