Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slmo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103624.2 | NP_001097094.1 | 2 | 1654 | 124 | |
NM_001144321.2 | NP_001137793.1 | 1 | 2007 | 124 | ||
NM_001298724.1 | NP_001285653.1 | 1 | 1235 | 124 | ||
NM_079966.3 | NP_524705.1 | 2 | 1116 | 124 | ||
NM_164677.2 | NP_723148.1 | 1 | 1235 | 124 | ||
GenBank mRNA | AY051484.1 | - | 1 | 1195 | 124 | |
BT029137.1 | - | 1 | 2002 | 124 | ||
BT099532.1 | - | 2 | 1115 | 124 |
Gene Symbol: CG34179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144322.2 | NP_001137794.1 | 1 | 2007 | 124 | |
NM_001298725.1 | NP_001285654.1 | 2 | 1654 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |