Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144330.2 | NP_001137802.1 | 3 - 4 | 662 | 100 | |
NM_078771.3 | NP_523495.2 | 2 - 3 | 997 | 100 | ||
NM_164702.2 | NP_723203.1 | 3 - 4 | 536 | 100 | ||
NM_164703.2 | NP_723204.1 | 3 - 4 | 523 | 100 | ||
NM_164704.2 | NP_723205.1 | 2 - 3 | 513 | 100 | ||
GenBank mRNA | AY069229.1 | - | 2 - 3 | 495 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |