Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nrv2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014475.2 | NP_001014475.1 | 3 - 4 | 713 | 62 | |
NM_001014476.2 | NP_001014476.1 | 4 - 5 | 519 | 62 | ||
NM_001273235.1 | NP_001260164.1 | 4 - 5 | 1205 | 62 | ||
NM_057820.4 | NP_477168.1 | 5 - 6 | 1238 | 62 | ||
NM_057821.4 | NP_477169.1 | 4 - 5 | 550 | 62 | ||
NM_164711.2 | NP_723216.1 | 4 - 5 | 726 | 62 | ||
GenBank mRNA | AY060289.1 | - | 3 - 4 | 700 | 62 | |
BT025048.1 | - | 4 - 5 | 517 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |