Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pvr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169439.2 | NP_001162910.1 | 13 - 14 | 3523 | 58 | |
NM_001169440.2 | NP_001162911.1 | 13 - 14 | 3523 | 58 | ||
NM_001201791.2 | NP_001188720.1 | 11 - 12 | 3292 | 58 | ||
NM_001201792.2 | NP_001188721.1 | 12 - 13 | 3192 | 58 | ||
NM_001273305.1 | NP_001260234.1 | 12 - 13 | 3496 | 58 | ||
NM_001273306.1 | NP_001260235.1 | 11 - 12 | 3289 | 58 | ||
NM_078785.4 | NP_523509.2 | 12 - 13 | 3319 | 58 | ||
NM_164801.3 | NP_723365.1 | 12 - 13 | 3319 | 58 | ||
NM_205925.3 | NP_995647.2 | 11 - 12 | 3289 | 58 | ||
NM_205926.3 | NP_995648.1 | 12 - 13 | 3354 | 58 | ||
NM_205927.3 | NP_995649.1 | 12 - 13 | 3936 | 58 | ||
GenBank mRNA | BT133154.1 | - | 13 - 14 | 3492 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |