Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_164833.2 | NP_723413.1 | 1 - 2 | 195 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |