Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp29Fb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144337.2 | NP_001137809.1 | 3 - 4 | 609 | 64 | |
NM_078792.3 | NP_523516.1 | 4 - 5 | 495 | 64 | ||
NM_164838.2 | NP_723426.1 | 4 - 5 | 538 | 64 | ||
GenBank mRNA | AF274012.1 | - | 4 - 5 | 495 | 64 | |
AY122216.1 | - | 4 - 5 | 495 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |