Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alien | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273347.1 | NP_001260276.1 | 4 - 5 | 974 | 80 | |
NM_001298842.1 | NP_001285771.1 | 4 - 5 | 974 | 80 | ||
NM_078793.6 | NP_523517.1 | 4 - 5 | 974 | 80 | ||
NM_164843.3 | NP_723438.1 | 4 - 5 | 974 | 80 | ||
GenBank mRNA | AF129079.1 | - | 4 - 5 | 934 | 80 | |
AY069836.1 | - | 4 - 5 | 927 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |