Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggamma30A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169446.3 | NP_001162917.2 | 1 - 2 | 109 | 90 | |
NM_001169447.2 | NP_001162918.1 | 2 - 3 | 279 | 90 | ||
NM_001201820.2 | NP_001188749.1 | 3 - 4 | 711 | 90 | ||
NM_001259021.2 | NP_001245950.1 | 3 - 4 | 730 | 90 | ||
NM_001298847.1 | NP_001285776.1 | 2 - 3 | 789 | 90 | ||
NM_080068.4 | NP_524807.1 | 3 - 4 | 593 | 90 | ||
NM_164851.3 | NP_723451.1 | 2 - 3 | 279 | 90 | ||
NM_164852.3 | NP_723452.1 | 2 - 3 | 789 | 90 | ||
GenBank mRNA | AJ250440.1 | - | 2 - 3 | 362 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |