Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bsk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169459.1 | NP_001162930.1 | 5 - 6 | 1521 | 96 | |
NM_001169461.3 | NP_001162932.1 | 5 - 6 | 1379 | 96 | ||
NM_164900.3 | NP_723541.1 | 6 - 7 | 1259 | 96 | ||
GenBank mRNA | AY070865.1 | - | 6 - 7 | 1136 | 96 | |
AY122221.1 | - | 6 - 7 | 1238 | 96 | ||
U49180.1 | - | 5 - 6 | 1522 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster Inventoried |