Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hoe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273130.1 | NP_001260059.1 | 5 - 6 | 1048 | 109 | |
NM_001273131.1 | NP_001260060.1 | 5 - 6 | 976 | 109 | ||
NM_001298678.1 | NP_001285607.1 | 4 - 5 | 775 | 109 | ||
NM_135031.4 | NP_608875.1 | 4 - 5 | 760 | 109 | ||
NM_135032.2 | NP_608876.1 | 4 - 5 | 693 | 109 | ||
NM_164603.2 | NP_723024.2 | 5 - 6 | 904 | 109 | ||
NM_175966.2 | NP_787980.1 | 5 - 6 | 988 | 109 | ||
NM_175967.2 | NP_787981.1 | 5 - 6 | 1026 | 109 | ||
NM_175968.2 | NP_787982.1 | 5 - 6 | 972 | 109 | ||
GenBank mRNA | AF145605.1 | - | 4 - 5 | 739 | 109 | |
AY050571.1 | - | 5 - 6 | 991 | 109 | ||
AY050572.1 | - | 4 - 5 | 742 | 109 | ||
BT016131.1 | - | 5 - 6 | 991 | 109 | ||
BT044574.1 | - | 4 - 5 | 688 | 109 | ||
BT126019.1 | - | 2 - 3 | 211 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |