Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104160.3 | NP_001097630.2 | 2 | 3791 | 77 | |
NM_001202194.1 | NP_001189123.1 | 2 | 3691 | 77 | ||
GenBank mRNA | BT132960.1 | - | 2 | 3691 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |