Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140802.3 | NP_649059.1 | 2 - 3 | 341 | 65 | |
GenBank mRNA | AY119557.1 | - | 2 - 3 | 320 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |