Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tey | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144509.3 | NP_001137981.1 | 4 - 5 | 1443 | 70 | |
NM_001275130.2 | NP_001262059.1 | 4 - 5 | 1413 | 70 | ||
NM_001275131.1 | NP_001262060.1 | 4 - 5 | 1258 | 70 | ||
NM_140883.2 | NP_649140.1 | 4 - 5 | 1443 | 70 | ||
GenBank mRNA | BT133456.1 | - | 4 - 5 | 1442 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |