Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RhoBTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275178.1 | NP_001262107.1 | 2 - 3 | 581 | 62 | |
NM_001275179.1 | NP_001262108.1 | 2 - 3 | 319 | 62 | ||
NM_140959.2 | NP_649216.2 | 2 - 3 | 581 | 62 | ||
GenBank mRNA | AF217287.1 | - | 2 - 3 | 558 | 62 | |
AY051710.1 | - | 2 - 3 | 548 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |