Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gli | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057254.4 | NP_476602.1 | 2 - 3 | 361 | 65 | |
NM_165127.2 | NP_723928.1 | 3 - 4 | 452 | 65 | ||
NM_165128.2 | NP_723929.1 | 2 - 3 | 241 | 65 | ||
NM_165129.2 | NP_723930.1 | 2 - 3 | 273 | 65 | ||
NM_165130.2 | NP_723931.2 | 2 - 3 | 734 | 65 | ||
GenBank mRNA | BT133203.1 | - | 2 - 3 | 358 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |