Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bchs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135124.2 | NP_608968.2 | 7 - 8 | 3574 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |