Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259010.2 | NP_001245939.1 | 2 - 3 | 514 | 74 | |
NM_135355.4 | NP_609199.1 | 2 - 3 | 514 | 74 | ||
GenBank mRNA | AY069492.1 | - | 2 - 3 | 486 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |