Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CadN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032106.2 | NP_001027277.1 | 18 - 19 | 8988 | 63 | |
NM_001032107.3 | NP_001027278.1 | 18 - 19 | 8988 | 63 | ||
NM_001032108.2 | NP_001027279.1 | 18 - 19 | 8985 | 63 | ||
NM_001032109.3 | NP_001027280.1 | 18 - 19 | 8985 | 63 | ||
NM_165224.2 | NP_724068.1 | 17 - 18 | 8973 | 63 | ||
NM_165225.2 | NP_724069.1 | 17 - 18 | 8973 | 63 | ||
NM_165226.2 | NP_724070.1 | 17 - 18 | 8976 | 63 | ||
NM_165227.3 | NP_724071.1 | 19 - 20 | 9132 | 63 | ||
NM_165228.2 | NP_724072.1 | 17 - 18 | 8973 | 63 | ||
NM_165229.2 | NP_724073.1 | 17 - 18 | 8973 | 63 | ||
NM_165230.2 | NP_724074.1 | 17 - 18 | 8976 | 63 | ||
NM_165231.3 | NP_724075.1 | 17 - 18 | 8976 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |