Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RtGEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103714.3 | NP_001097184.2 | 8 - 9 | 4327 | 84 | |
NM_057826.5 | NP_477174.1 | 7 - 8 | 2379 | 84 | ||
NM_165327.3 | NP_724258.1 | 7 - 8 | 2019 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |